Learn More
Apexbio Technology LLC 3-Deazaadenosine hydrochloride(Synonyms: 3-Deazaadenosine HCl, C3-Deazaadenosine hydrochloride, 3-Deaza-adenosine hydrochloride), 1mg, CAS: 86583-19-9.
SDP
Supplier:  Apexbio Technology LLC B84701
Encompass_Preferred
3-Deazaadenosine hydrochloride (CAS 86583-19-9) is a selective inhibitor of S-adenosylhomocysteine hydrolase (SAHH) exhibiting inhibition with a Ki value around 3 9 M It disrupts SAHH-dependent methyl metabolism pathways thereby altering intracellular methyltransferase reactions Due to this biochemical activity it serves as a research agent for studying inflammation cell proliferation and HIV infection as well as investigations into enzyme-mediated methylation processes and related pathological mechanisms
Catalog No. 50-000-60498