Learn More
Apexbio Technology LLC 3-Deazaneplanocin A (DZNep) hydrochloride, 25mg
SDP
Supplier:  Apexbio Technology LLC A818225
Encompass_Preferred
3-Deazaneplanocin A (DZNep) hydrochloride is a selective inhibitor of ENZ2 inhibitor with IC50 value of 0.08-0.24 M [1].ENZ2 is a sub-unit of PRC2 and plays an important role in regulating cell proliferation and cell-cycle progression. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5071