Learn More
Cayman Chemical 3-DeazaurIdIn 10mg
SDP
Supplier:  Cayman Chemical 2312510 MG
Encompass_Preferred
A nucleoside analog; converted intracellularly to 3-deazauridine triphosphate, which inhibits the biosynthesis of CTP; inhibits the growth of L1210 leukemia cells when used at a concentration of 6 µM; dose-dependently reduces mortality in a mouse model of leukemia; enhances the incorporation of decitabine into DNA in leukemia cells when used at a concentration of 20 µM; reduces mortality when used at a dose of 100 or 150 mg/kg and combined with decitabine in a mouse model of leukemia
Catalog No. 50-299-3901