Learn More
Cayman Chemical 3-Deoxy-3-fluorogunosIn 10mg
SDP
Supplier:  Cayman Chemical 3569910 MG
Encompass_Preferred
A nucleoside analog an inhibitor of HCV NS5B (IC50 = 1.8 µM)
Catalog No. 50-291-1232