Learn More
Medchemexpress LLC 3-deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) | 153666-19-4 | 99.9% | C9H16O9 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14553310MG
Encompass_Preferred
3-Deoxy-D-glycero-D-galacto-2-nonulosonic acid (KDN) is a deaminated sialic acid derivative provided as a white to off-white solid for biochemical research. The compound offers high purity and reliable aqueous solubility for studies in glycobiology, sialylation, and related analytical workflows.
  • High purity (99.9%) suitable for analytical and biochemical applications.
  • Water soluble at up to 100 mg/mL; ultrasonic assistance may be required.
  • Solid form with defined storage stability under recommended conditions.
  • Molecular weight 268.22 g/mol and formula C9H16O9 for precise mass calculations.
  • Relevant endogenous metabolite for studies of sialic acid biology.
Catalog No. 50-002-61136