Learn More
Cayman Chemical 3-deoxy Galactoson 10mg
SDP
Supplier:  Cayman Chemical 1680110 MG
Encompass_Preferred
A 1,2-dicarbonyl compound originating from the degradation of galactose; can be formed by Maillard or caramelization reactions
Catalog No. 50-296-6433