Learn More
Cayman Chemical 3-DeoxygunosIn 10mg
SDP
Supplier:  Cayman Chemical 1790710 MG
Encompass_Preferred
A ligand that can be complexed with enzymes, such as purine nucleoside phosphorylase, and receptors, such as G-coupled receptors, in order to study structure-activity relationships; also used to selectively impair transcription
Catalog No. 50-296-6128