Learn More
Medchemexpress LLC 3-desacetyl cefotaxime lactone | 66340-33-8 | MFCD24386610 | 97.0% | C14H13N5O5S2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13539410MG
Encompass_Preferred
3-Desacetyl cefotaxime lactone (CAS 66340-33-8) is the lactone form of the active metabolite of the third-generation cephalosporin antibiotic cefotaxime. It is provided as a white to off-white solid and is intended for use as an analytical reference standard, impurity characterization, and metabolic or degradation studies in pharmaceutical research.
  • High purity (97.0%) suitable for analytical applications.
  • White to off-white solid, amenable to standard handling and storage.
  • Useful as an impurity and metabolite standard for cephalosporin studies.
  • Supplied in small-mass quantities appropriate for reference use.
  • Supports structural characterization and stability testing.
Catalog No. 50-002-59351