Learn More
Medchemexpress LLC 3-epideoxycholic acid | 570-63-8 | >98.0% | 10 MG

Supplier: Medchemexpress LLC HY4881410MG
3-Epideoxycholic acid is a secondary bile acid and microbial metabolite of deoxycholic acid used as a research reagent and FXR modulator in biochemical and pharmacological studies. It is supplied as a high-purity analytical standard suitable for receptor modulation assays, metabolite identification, and analytical chemistry.
- High purity: >98.0%.
- Molecular formula C24H40O4; molecular weight 392.6 g·mol⁻¹.
- Used as an FXR modulator and endogenous metabolite standard.
- Suitable for analytical, biochemical, and pharmacological research.
- Available in small pack sizes for analytical use, e.g., 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.