Learn More
Medchemexpress LLC 3-ethoxybenzoic acid | 621-51-2 | MFCD00016517 | 98.7% | 166.18 g/mol | C9H10O3 | 500 MG

Supplier: Medchemexpress LLC HYW048341500MG
3-ethoxybenzoic acid is a substituted benzoic acid used as a raw material and intermediate in organic synthesis. It is provided for laboratory research and is available in solid and solution formats.
- Chemical formula: C9H10O3.
- Molecular weight: 166.18 g/mol.
- Cas number: 621-51-2.
- Purity: 98.74% (page lists 97.0% at summary; batch-specific purity shown in documentation).
- Available formats: 500 mg solid, 1 g, 5 g, 10 g, 50 g (quote), and 10 mM (1 mL) solution in DMSO.
- Storage: solid at room temperature (stable up to 3 years); in solvent store at -80°C or -20°C per documentation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.