Learn More
Medchemexpress LLC 3-(ethoxycarbonyl)thiazolidine-4-carboxylic acid | 127657-29-8 | MFCD04117921 | 98.0% | 205.24 g/mol | C7H11NO4S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10828510MM 1ML
Encompass_Preferred
(Rac)-Telmesteine is a racemic thiazolidine-4-carboxylic acid protease inhibitor supplied as a ready-to-use solution in DMSO. The compound is reported at high purity and is intended for research use as an enzyme stabilizer and reference compound in applications that include protease-containing detergents and cleaning agents.
  • Protease inhibitor and enzyme stabilizer
  • Ready-to-use 10 mM solution in DMSO
  • High purity suitable for research (98.0%)
  • Small-volume format for screening and assay work
  • Compatible with protease-containing detergents and cleaning agents
Catalog No. 50-002-43435