Learn More
ACCUTEC BLADES INC (3P) 3 Facet Single Edge Stainless Steel Razor blade
Supplier:  ACCUTEC BLADES INC (3P) 6201790000
Encompass_Preferred
Industrial Razor Blade GEM 3 Facet, Single Edge, Stainless Steel BLADE, ACCUFORGE CLAMSHELL S/STL .009" UNCOATED Aluminum back (100/BX)
Catalog No. NC3721093