Learn More
Medchemexpress LLC 3-feruloylquinic acid | 1899-29-2 | 98.5% | 368.34 g/mol | C17H20O9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN659910MG
Encompass_Preferred
3-Feruloylquinic acid is a chlorogenic-acid-type derivative of quinic acid-bound phenolic acid. It exhibits antioxidant and anti-inflammatory activities and is supplied for research use only. Molecular formula C17H20O9; molecular weight 368.34 g/mol; CAS 1899-29-2.
  • Shows antioxidant and anti-inflammatory activity.
  • Supplied at high purity (>98%).
  • Available in small research sizes (1 mg, 5 mg, 10 mg; larger sizes by quote).
  • Documentation available: data sheet, certificate of analysis, SDS, H-NMR, and RP-HPLC.
  • Intended for research use only; not for human or therapeutic use.
Catalog No. 50-002-89934