Learn More
Medchemexpress LLC (3-Fluoro-2-formylphenyl)boronic acid | 871126-15-7 | 167.93 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00099410G
Encompass_Preferred
(3-Fluoro-2-formylphenyl)boronic acid is a biochemical reagent used in life science research. It is categorized as biochemical assay reagents and undergoes unprecedented tautomeric rearrangement in solution. This product is for research use only and has not been fully validated for medical applications.
  • Can be used as a biological material or organic compound
  • Undergoes tautomeric rearrangement in solution
  • Suitable for life science related research
  • Recommended storage at 4°C under nitrogen
  • In solvent storage at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-47118