Learn More
Medchemexpress LLC (3-Fluoro-2-formylphenyl)boronic acid | 871126-15-7 | 167.93 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW000994
Encompass_Preferred
(3-Fluoro-2-formylphenyl)boronic acid is a biochemical reagent used as a biological material or organic compound for life science related research. It undergoes unprecedented tautomeric rearrangement in solution.
  • Stable under recommended storage conditions
  • Recommended storage at 4°C under nitrogen
  • Suitable for laboratory chemicals and substance manufacturing
Catalog No. 50-004-50025