Learn More
Medchemexpress LLC 3-Fluoro-4-methoxybenzaldehyde | 351-54-2 | 154.14 | 5 G

Supplier: Medchemexpress LLC HYW0076145G
3-Fluoro-4-methoxybenzaldehyde (3-Fluoro-para-anisaldehyde) is a drug intermediate used in the synthesis of fluorinated derivatives of Combretastatin A-4, which possesses anti-cancer activity. It is for research use only.
- Can be used to synthesize fluorinated derivative of Combretastatin A-4
- Possesses anti-cancer activity
- For research use only
- Molecular weight: 154.14
- Formula: C8H7FO2
- Appearance: Liquid
- Color: Light yellow to yellow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.