Learn More
Medchemexpress LLC 3-fluoro-4-phenylbenzoic acid | 137045-30-8 | MFCD05664334 | 216.21 | C13H9FO2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW014136500MG
Encompass_Preferred
Flurbiprofen impurity 3 is a research-grade impurity of the NSAID flurbiprofen supplied as an analytical standard for impurity profiling, method development, and reference studies. The substance is a solid with molecular formula C13H9FO2, molecular weight 216.21 Da, and CAS 137045-30-8. Store under the conditions specified in the certificate of analysis.
  • Designed for impurity profiling and reference standard use.
  • Suitable for analytical method development such as HPLC and LC-MS.
  • Molecular formula C13H9FO2, molecular weight 216.21 Da, CAS 137045-30-8.
  • Solid form; follow the certificate of analysis for storage and handling.
  • Supplied in laboratory-scale quantities for analytical applications.
Catalog No. 50-003-04072