Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-fluoro-N-[(E)-(5-pyridin-2-ylsulfanylfuran-2-yl)methylideneamino]benzamide | 352336-36-8 | 99.4% | C17H12FN3O2S | 10MG
SDP

Supplier:  Medchemexpress LLC HY13351110MG

Encompass_Preferred

HY-133511 (MLS000544460) is a selective, reversible inhibitor of Eya2 phosphatase used in biochemical and cellular research. It is supplied as a white to off-white solid (C17H12FN3O2S) with high reported purity and characterized potency for use in mechanism and preclinical studies.

  • selective, reversible inhibition of Eya2 phosphatase
  • high reported purity (99.42%) suitable for research assays
  • characterized potency: Kd 2.0 μM and IC50 4 μM
  • high solubility in DMSO (125 mg/mL) and formulatable for animal dosing
  • powder storage stability: -20°C for up to 3 years
  • white to off-white solid with molecular weight 341.36 g/mol

Catalog No. 50-002-58367


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.