Learn More
Medchemexpress LLC 3-fluorobutan-1-amine hydrochloride | 1780799-10-1 | 95.0% | 127.59 g·mol-1 | C4H11ClFN | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0497701G
Encompass_Preferred
3-fluorobutan-1-amine hydrochloride is a fluorinated primary amine used as a synthetic intermediate in medicinal chemistry research. It is supplied as an off-white to light yellow solid, with molecular formula C4H11ClFN, molecular weight 127.59 g·mol-1, and typical purity around 95.0%. Storage recommendations: keep sealed at room temperature and protected from moisture and light; in solvent, store at -80°C for long-term preservation.
Catalog No. 50-002-10535