Learn More
Medchemexpress LLC 3-Formylsalicylic acid | 610-04-8 | 166.13 | 250 MG

Supplier: Medchemexpress LLC 250MGHY41570
3-Formylsalicylic acid is a noncompetitive estrone sulfatase inhibitor, exhibiting an IC50 value of 0.15 mM and a Ki value of 0.12 mM. This compound demonstrates low acute toxicity.
- Acts as a noncompetitive estrone sulfatase inhibitor
- Shows low acute toxicity
- Targets Estrogen Receptor/ERR
- Involves the Vitamin D Related/Nuclear Receptor pathway
- Appears as a solid
- Melting point of 177.5 °C
- Boiling point of 306.9 °C at 760 mmHg
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.