Learn More
Medchemexpress LLC 3-Formylsalicylic acid | 610-04-8 | 98.5% | 166.13 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY41570100MG
Encompass_Preferred
3-Formylsalicylic acid is a noncompetitive estrone sulfatase inhibitor. It exhibits an IC50 value of 0.15 mM and a Ki value of 0.12 mM, and has demonstrated low acute toxicity. This product is for research use only.
  • Noncompetitive estrone sulfatase inhibitor
  • IC50 value of 0.15 mM
  • Ki value of 0.12 mM
  • Low acute toxicity
Catalog No. 50-004-03661