Learn More
Medchemexpress LLC 3-Formylsalicylic acid | 610-04-8 | MFCD00143165 | 98.5% | 166.13 | 500 MG

Supplier: Medchemexpress LLC HY41570500MG
3-Formylsalicylic acid is a hydroxybenzoic acid, an aldehyde, and a phenol derived from salicylic acid. It is identified as an irritant.
- Molecular formula C8H6O4
- Molecular weight 166.13 g/mol
- Harmful if swallowed
- Causes serious eye irritation
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.