Learn More
Medchemexpress LLC 3-formylsalicylic acid | 610-04-8 | MFCD00143165 | 98.5% | 166.13 | C8H6O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY4157010MG
Encompass_Preferred
3-Formylsalicylic acid is a small-molecule, noncompetitive estrone sulfatase inhibitor used in biochemical and pharmacological research to study estrogen metabolism and related pathways. The compound is characterized by defined potency and high purity for in vitro studies.
  • Noncompetitive estrone sulfatase inhibitor (IC50 0.15 mM; Ki 0.12 mM).
  • High purity for research use (~98.5%).
  • Molecular weight 166.13 and formula C8H6O4.
  • Provided in small research pack sizes suitable for assay screening.
  • Reported low acute toxicity in available literature.
Catalog No. 50-002-97860