Learn More
Medchemexpress LLC 3-(glycidyloxy)propyltrimethoxysilane | 2530-83-8 | MFCD00005144 | 99.4% | 236.34 | C9H20O5Si | 100 G
SDP
Supplier:  Medchemexpress LLC HYW089777100G
Encompass_Preferred
3-Glycidyloxypropyltrimethoxysilane is an epoxy-functional organosilane used to introduce glycidoxy groups onto surfaces and polymer matrices for improved adhesion, crosslinking, and surface modification.
  • Epoxy-functional silane coupling agent suitable for surface modification and adhesion promotion.
  • Used as a crosslinking and adhesion promoter in coatings, composites, and sol-gel systems.
  • Compatible with inorganic substrates and organic polymers to improve interfacial bonding.
  • High purity suitable for research and formulation (purity: 99.4%).
  • Available in multiple packaging sizes for lab-scale and larger syntheses.
Catalog No. 50-002-08679