Learn More
Cayman Chemical 3-GunIdInopropIonIc AcId 500mg
SDP
Supplier:  Cayman Chemical 16725500 MG
Encompass_Preferred
A creatine analog that alters skeletal muscle energy expenditure; reduces cellular ATP, creatine, and phosphocreatine levels and stimulates AMPK, activating PGC-1α; increases the expression of genes for oxidative phosphorylation, electron transport chain, and mitochondrial biogenesis
Catalog No. 50-296-5371