Learn More
Medchemexpress LLC 3-hydroxy-1,2,3-benzotriazin-4(3H)-one | 28230-32-2 | MFCD00042803 | 99.9% | 163.13 g/mol | C7H5N3O2 | 100g
SDP
Supplier:  Medchemexpress LLC HYY0493100G
Encompass_Preferred
HODHBt (HOOBt) inhibits STAT5-SUMO interaction by blocking SUMOylation of phosphorylated STAT5 HODHBt enhances the magnitude of IL-15 signaling and significantly increases the natural killer (NK) cell cytotoxicity phenotype and function and the generation of cytokine-induced memory-like (CIML) natural killer (NK) cells HODHBt can be used for research of HIV-infection and cancer[1]
Catalog No. 50-002-69966