Learn More
Medchemexpress LLC 3-hydroxy-2-phenylpropanoic acid | 552-63-6 | MFCD00004255 | 99.5% | 166.18 g/mol | C9H10O3 | 10 G

Supplier: Medchemexpress LLC HYW04119410G
Tropic acid (DL-tropic acid) is an organic small molecule used as a pharmaceutical intermediate, for example in the synthesis of atropine. Intended for research use only; not for human or veterinary use.
- Used as a pharmaceutical intermediate.
- Cas number 552-63-6.
- Molecular formula C9H10O3, molecular weight 166.18 g/mol.
- High purity (approx. 99.5%).
- Available in solid pack sizes from 100 mg to 50 g, including 10 G.
- Supplied with datasheet and certificate of analysis for verification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.