Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Hydroxy-4-nitrobenzoic acid | 619-14-7 | ≥98.0% | C7H5NO5 | 50 G
SDP

Supplier:  Medchemexpress LLC HY6934150G

Encompass_Preferred

3-Hydroxy-4-nitrobenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.

  • Biochemical assay reagents
  • CAS number: 619-14-7
  • Molecular formula: C7H5NO5
  • Molecular weight: 183.12
  • Purity: ≥98.0%
  • Store as powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-34293


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.