Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 3-Hydroxy-7-methoxyflavone | 7478-60-6 | 99.4% | 268.26 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN92415MG

Encompass_Preferred

3-Hydroxy-7-methoxyflavone is a flavonoid isolated from natural Salvia elegans, known for its anti-anxiety effects. This compound is for research use only.

  • Isolated from natural Salvia elegans.
  • Exhibits anti-anxiety effects.
  • Cytotoxicity against human KB cells with an ED50 of 26 µg/mL.
  • Recommended storage at room temperature for 3 years, or in solvent at -80°C for 2 years or -20°C for 1 year.
  • Soluble in DMSO at 50 mg/mL.

Catalog No. 50-004-26714


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.