Learn More
Cayman Chemical 3-hydroxy Carbofurn 10mg
SDP
Supplier:  Cayman Chemical 2414610 MG
Encompass_Preferred
An active metabolite of carbofuran formed in plants, insects, and mammals; LC50 = 0.75 ppm in C. pipiens; LD50 = >500 mg/kg in M. domestica; considered an environmental toxin
Catalog No. 50-293-3374