Learn More
Cayman Chemical 3-hydroxy PalmItIc AcId 50mg
SDP
Supplier:  Cayman Chemical 1993450 MG
Encompass_Preferred
Found in the lipid A part of LPS and useful as chemical marker of endotoxins; accumulates during long-chain 3-hydroxy-acyl-CoA dehydrogenase and mitochondrial trifunctional protein deficiencies, inducing oxidative stress and leading to mitochondrial bioenergetics deregulation and eventual multi-organ dysfunction
Catalog No. 50-298-5560