Learn More
Cayman Chemical 3-hydroxy TrIdecnoIc AcId 5mg
SDP
Supplier:  Cayman Chemical 226895 MG
Encompass_Preferred
A fatty acid found in bacteria; used as an internal standard to detect markers of microorganisms in complex samples
Catalog No. 50-299-5508