Learn More
Medchemexpress LLC 3-Hydroxybenzoic acid | 99-06-9 | 138.12 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0040495G
Encompass_Preferred
3-Hydroxybenzoic acid (Standard) is an analytical standard intended for research and analytical applications. It is an orally active endogenous metabolite.
  • Agonist for GPR81 and GPR109A
  • Acts as stress response desensitizer
  • Can be used for anti-inflammatory and analgesic studies
Catalog No. 50-004-45761