Learn More
Medchemexpress LLC 3-Hydroxybutyric acid sodium | 150-83-4 | 98.0% | 126.09 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01045210G
Encompass_Preferred
3-Hydroxybutyric acid sodium (β-Hydroxybutyric acid sodium) is a metabolite that is elevated in type I diabetes. It can modulate the properties of membrane lipids and is capable of interacting with lipids (modeled using a DPPC monolayer) and altering phase behavior at clinical concentrations. It also diminishes the interfacial viscosity of DPPC Monolayers.
  • Modulates properties of membrane lipids
  • Diminishes interfacial viscosity of DPPC Monolayers
Catalog No. 50-004-53897