Learn More
Medchemexpress LLC 3'-hydroxyflavanone | 92496-65-6 | MFCD00017738 | >98.0% | 240.25 g/mol | C15H12O3 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW2852431G
Encompass_Preferred
3'-Hydroxyflavanone is a cyclized flavanone (C15H12O3) reported to induce apoptosis in HeLa cells and to possess antitumor activity in cellular studies. It is employed in biochemical and pharmacological research to probe apoptosis pathways and to study flavonoid bioactivity.
  • Cyclized flavanone scaffold with a 3'-hydroxy substitution.
  • Reported to induce apoptosis in HeLa cells.
  • Demonstrated antitumor activity in preclinical cellular studies.
  • Useful as a biochemical probe for apoptosis-related pathways.
  • Supplied as a solid with defined molecular formula and molecular weight.
Catalog No. 50-002-80540