Learn More
Medchemexpress LLC 3-hydroxyglutaric acid | 638-18-6 | MFCD06208235 | 99.9% | 148.11 g/mol | C5H8O5 | 5 MG

Supplier: Medchemexpress LLC HY1134115MG
3-Hydroxyglutaric acid is a glutaric acid derivative supplied as a high-purity analytical standard for research and analytical applications. Provided as a white to off-white solid in small packaged quantities, it is suitable for biochemical assays, metabolite reference, and analytical method development when handled and stored per manufacturer recommendations.
- High purity: 99.93%.
- Molecular weight 148.11 g/mol and formula C5H8O5.
- Available in small solid pack sizes from 5 mg to 100 mg.
- Water soluble (250 mg/mL); PBS solubility 100 mg/mL; may require ultrasonic treatment.
- Stable when stored as powder at recommended temperatures (powder: -20°C for 3 years, 4°C for 2 years).
- Documentation available: datasheet, COA, SDS, and spectral reports.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.