Learn More
Medchemexpress LLC (3-Hydroxymethyl)phenylboronic acid | 87199-15-3 | MFCD01317846 | 151.96 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00741810G
Encompass_Preferred
3-(Hydroxymethyl)phenylboronic acid is a biochemical reagent suitable for life science-related research as a biological material or organic compound.
  • Used as a reactant for copper-mediated trifluoromethylation, copper-catalyzed transformations from arylboronic acids in water, Mitsunobu, Suzuki, and amidation reactions with hydroxyphenylamino bromopyrazinecarboxylate.
  • Involved in the synthesis of biologically active molecules such as Mycobacterium tuberculosis H37Rv chorismate mutase inhibitors via Suzuki coupling reactions.
  • Involved in the synthesis of HIV protease inhibitors with antiviral activity against drug-resistant viruses.
  • Involved in the synthesis of pyrrole derivatives for use as PDE4B inhibitors.
Catalog No. 50-004-60317