Learn More
Medchemexpress LLC 3-Hydroxyoctanoic acid | 14292-27-4 | 98.0% | 160.21 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11305810MG
Encompass_Preferred
3-Hydroxyoctanoic acid is a hydroxylated fatty acid found in the LPS of *Pseudomonas aeruginosa* and in methyl-branched poly(3-hydroxyalkanoate) (PHA) polymers produced by *Pseudomonas oleophores*. It acts as an agonist of the orphan receptor GPR109B, increasing intracellular calcium in human neutrophils endogenously expressing GPR109B.
  • Prevents lipolysis in human adipocytes.
  • Upregulated in human plasma in response to a ketogenic diet.
  • Plasma levels increased 3.41-fold in human male runners exhausted on a treadmill.
  • Increased in a mouse model of autism spectrum disorder (ASD) fed a high-glycemic diet.
Catalog No. 50-004-36133