Learn More
Selleck Chemical LLC 3-Hydroxypicolinic acid
SDP
Supplier:  Selleck Chemical LLC S625225MG
Encompass_Preferred
3-Hydroxy picolinic acid is a picolinic acid derivative and is a member of the pyridine family It is used as a matrix for nucleotides in MALDI mass spectrometry analyses
Catalog No. 50-000-03883