Learn More
Cayman Chemical 3-HydroxypIcolInIc AcId 1g
SDP
Supplier:  Cayman Chemical 345301 G
Encompass_Preferred
A matrix substance for MALDI-MS; has been used in the analysis of oligonucleotides and proteins; has been used as a complexation agent for metal complexes; is an intermediate in the biosynthesis of virginiamycin S1
Catalog No. 50-295-0571