Learn More
Medchemexpress LLC 3-hydroxypyridine | 109-00-2 | MFCD00006378 | 95.10 g/mol | C5H5NO | 100g
SDP
Supplier:  Medchemexpress LLC HYY1129100G
Encompass_Preferred
3-Hydroxypyridine is a compound that can be isolated from bamboo grass As an endogenous photosensitizer present in human skin 3-hydroxypyridine can mediate oxidative stress proliferation inhibition and apoptosis of skin cells through UVA/UVB excitation[1][2]
Catalog No. 50-002-67478