Learn More
Medchemexpress LLC 3-Hydroxyvaleric acid | 10237-77-1 | 98.0% | 118.13 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11300410MM/1ML
Encompass_Preferred
3-Hydroxyvaleric acid is a five-carbon ketone body byproduct that can undergo an anaplerotic reaction to replenish the intermediates of the tricarboxylic acid cycle. It is also an important monomer for the synthesis of polyhydroxyalkanoates (PHA), a type of biodegradable plastic used in environmentally friendly packaging, disposable products, and medical materials.
  • Potentially brain penetrant.
  • Rapidly metabolized to TCA cycle precursors.
  • Can derive the neurotransmitters glutamate and GABA, potentially alleviating diseases or conditions characterized by reduced or net brain carbon deficiency, such as glucose transporter type I deficiency syndrome (G1D).
Catalog No. 50-004-39767