Learn More
Medchemexpress LLC 3-Hydroxyxanthone (3-Hydroxy-xanthen-9-one) | 3722-51-8 | 98.4% | 212.20 | 50 G
Supplier: Medchemexpress LLC HYW01355750G
3-Hydroxyxanthone is a xanthone compound. It inhibits NADPH-catalysed lipid peroxidation in human umbilical vein endothelial cells (HUVECs) and also inhibits TNF-alpha induced ICAM-1 expression.