Learn More
Medchemexpress LLC 3-Indoleacetic acid | 87-51-4 | MFCD00005636 | 99.9% | 175.19 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1856910MM/1ML
Encompass_Preferred
3-Indoleacetic acid is an IAA hormone and growth regulator that promotes plant nutritional growth through processes like cell expansion, differentiation, morphogenesis, and organogenesis. For research use only.
  • IAA hormone and growth regulator
  • Promotes plant nutritional growth through cell expansion, differentiation, morphogenesis, and organogenesis
Catalog No. 50-003-87932