Learn More
Medchemexpress LLC 3-Indoleacetic acid-d5 | 76937-78-5 | 99.76% | 180.21 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY18569S5MG
Encompass_Preferred
3-Indoleacetic acid-d5 is the deuterium labeled 3-Indoleacetic acid (HY-18569). It can be used as an internal standard for the assay of IAA releases by alkaline hydrolysis of ester and amide conjugates.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-87862