Learn More
Medchemexpress LLC 3-indoleacetic acid-d5 | 76937-78-5 | MFCD00209745 | 99.8% | 180.21 | C10H4D5NO2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY18569S10MG
Encompass_Preferred
3-Indoleacetic acid-d5 is a deuterium-labeled analogue of indole-3-acetic acid used primarily as an internal standard for quantitative and mass-spectrometry assays. It is supplied as a stable, high-purity powder suitable for preparing calibration standards and spiking biological samples. Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C.
  • Deuterium-labeled internal standard for IAA quantification.
  • High isotopic and chemical purity for reliable mass-spec performance.
  • Stable powder form for accurate standard preparation.
  • Suitable for LC-MS/MS and GC-MS assay workflows.
  • Documented stability and storage conditions for reproducible results.
Catalog No. 50-002-97958