Learn More
Medchemexpress LLC 3-Indoleacetic acid, suitable for plant cell culture | 87-51-4 | 99.8% | 175.18 | 5 G

Supplier: Medchemexpress LLC HY18569B5G
3-Indoleacetic acid, suitable for plant cell culture (IAA), is a naturally occurring plant growth hormone widely present in plants, bacteria, and fungi. It not only promotes plant growth but also protects bacteria from toxic damage to cell membrane potential caused by other indole intermediate products. This product can be used in plant cell culture.
- Naturally occurring plant growth hormone
- Widely present in plants, bacteria, and fungi
- Promotes plant growth
- Protects bacteria from toxic damage to cell membrane potential caused by other indole intermediate products
- Can be used in plant cell culture
- Appearance: solid
- Color: white to off-white
- Shipping: room temperature in continental US
- Storage: powder -20°C 3 years; in solvent -80°C 6 months, -20°C 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.