Learn More
Apexbio Technology LLC 3-Indoleacetonitrile 771-51-7 1g
SDP
Supplier:  Apexbio Technology LLC C71661000
Encompass_Preferred
3-Indoleacetonitrile (CAS 771-51-7) is an indole-derived small molecule produced through the metabolic pathway of tryptophan It acts as a metabolic intermediate in the biosynthesis of indole-3-acetic acid (IAA) a key phytohormone involved in plant growth and developmental regulation In biomedical research 3-Indoleacetonitrile is employed to investigate mechanisms of tryptophan metabolism and its downstream effects on cellular growth developmental signaling cascades and the pathophysiology of metabolic disorders Its application facilitates the study of indole compound functions in both plant biology and broader metabolic contexts
Catalog No. 50-000-90058