Learn More
Selleck Chemical LLC 3-Indolepropionic acid S4809-25mg
SDP
Supplier:  Selleck Chemical LLC S480925MG
Encompass_Preferred
3-Indolepropionic acid (IPA indole-3-propionic acid) a gut microbial metabolite is a potent neuroprotective antioxidant and plant auxin
Catalog No. 50-311-1972