Learn More
Medchemexpress LLC 3-indolepropionic acid | 830-96-6 | MFCD00005660 | 99.9% | 189.21 g/mol | C11H11NO2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW015229R50MG
Encompass_Preferred
3-Indolepropionic acid is an indole-derived metabolite and antioxidant used as an analytical standard and research reagent. It is commonly applied in biochemical and neuroprotective research, and is suitable for analytical workflows requiring a stable, well-characterized reference compound.
  • Indole-derived antioxidant and endogenous metabolite.
  • High purity suitable for analytical and research use.
  • Supplied as a small-scale standard for assay calibration and method development.
  • Compatible with HPLC, LC-MS, and spectroscopic analyses.
  • Applicable to studies of neuroprotection and oxidative stress.
Catalog No. 50-003-02649